Orders LinkContactUsLinkComponent
EasyCurve™ Mop System, Contec®
EasyCurve™ Mop System, Contec®
EasyCurve™ Sterile Ribbed Microfiber Mop Head, 36×11×2 cm (14×1.25×0.75")
Catalog # 10153-826
Supplier:   Contec
Supplier #:  EZMP0350
EasyCurve™ Mop System, Contec®
Catalog # 10153-826
Supplier:  Contec
EasyCurve™ Mop System, Contec®
EasyCurve™ Mop System, Contec®
EasyCurve™ Sterile Ribbed Microfiber Mop Head, 36×11×2 cm (14×1.25×0.75")
Price
$1,390.00
Log in for availability
Case of 48
Product Details & Documents
The EasyCurve™ Mop was designed specifically for use in critical and controlled environments. It is now available with a ribbed microfiber face, ensuring smooth and easy mopping with improved particle pick-up and contamination removal.
  • All components are autoclavable; ideal for a wide range of cleaning applications in the most critical sterile environments
  • Lightweight mop head and handle significantly reduces operator fatigue
  • Single-use mop is economical and easy to install without tools
  • Curved mop and pivoting mop frame provide effortless contact with surface
  • Ribbed microfiber face provides better contaminant removal
The combination of the curved mop face and the pivoting connector results in effortless surface contact and outstanding maneuverability. This makes the EasyCurve™ an ideal cleaning tool for ceilings, walls, and floors. All components of the EasyCurve™ Mop are autoclavable. Available in both nonsterile and validated sterile versions.
Specifications
  • Description:
    EasyCurve™ Sterile Ribbed Microfiber Mop Head
  • Size:
    36×11×2 cm (14×1.25×0.75")
Accessories
Product Information
  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ anodized aluminum telescopic handle
    QuickConnect™ anodized aluminum telescopic handle
    33502-715
    127 - 234 cm (50 - 92")
    Log in for availability
    Each - $204.00
Product Information
  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ stainless steel telescopic handle
    QuickConnect™ stainless steel telescopic handle
    33502-716
    41 - 76 cm (16 - 30")
    Log in for availability
    Each - $385.00
Product Information
  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ anodized aluminum telescopic handle
    QuickConnect™ anodized aluminum telescopic handle
    33502-714
    41 - 76 cm (16 - 30")
    Log in for availability
    Each - $180.00
Product Information
  • Catalog #
    Availability
    Price
  • Stainless Steel Connecting Kit for Multi-Bucket System
    Stainless Steel Connecting Kit for Multi-Bucket System
    89073-986
    Log in for availability
    Each - $97.10
Product Information
  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ stainless steel telescopic handle
    QuickConnect™ stainless steel telescopic handle
    33502-717
    127 - 234 cm (50 - 92")
    Log in for availability
    Each - $561.00