Advansta Blot Development Folders
Blot Development Folders, 5 x 7"
Catalog # 490016-806
Supplier: Advansta
Supplier #: L-07020-025
Jump to:
Product Details & Documents
Advansta's blot development folders are transparent plastic folders that hold your blot while it is exposed to film or a CCD camera. These folders keep your blot flat for perfect film exposures, avoiding the wrinkling or bunching that can occur with plastic wrap.
- Clear images
- Suitable for all blots
- Safe blot handling
- Convenient size
Development folders are suitable for chemiluminescence and fluorescence imaging. You can move your blot without scratching of putting pressure on the surface. Folders allow you to image up to two blots imultaneously in one folder.
Specifications
- Description:Blot Development Folders
- Dimensions:5 x 7"