Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Ammonia solution strong NF, ACS

Ammonia solution strong NF, ACS

Drum, steel/PE outer, 385 lbs.
Catalog # JT3226-20
Corrosive
Irritant
Environment
Supplier #:  3226-20
Ammonia solution strong NF, ACS
Catalog # JT3226-20
CAS Number:  1336-21-6
Ammonia solution strong NF, ACS
Ammonia solution strong NF, ACS

Drum, steel/PE outer, 385 lbs.

CAS Number:  1336-21-6
MOC:  Regulated
Price
$3,569.49
Log in for availability
Each (385lb)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Ammonia Solution, Strong; Ammonium Hydroxide

Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • ADR:8,III
  •  :Management of Change (MOC) category = R
  • UN:2672

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • NF - Identification:
    Passes Test
  • NF - Nonvolatile Residue:
    ≤0.02 %
  • NF - Readily Oxidizable Substances:
    Passes Test
  • NF - Assay (NH₃)(NF):
    27.0 - 30.0 %
  • ACS - Assay (as NH₃):
    28.0 - 30.0 %
  • ACS - Carbon Dioxide (CO₂):
    ≤0.002 %
  • ACS - Residue after Ignition:
    ≤0.002 %
  • ACS - Substances Reducing Permanganate:
    Passes Test
  • Appearance:
    Passes Test
  • Arsenic and Antimony (as As):
    ≤3 ppm
  • Chloride (Cl):
    ≤0.5 ppm
  • Trace Impurities - Heavy Metals:
    ≤0.5 ppm
  • Nitrate (NO₃):
    ≤2 ppm
  • Phosphate (PO₄):
    ≤2 ppm
  • Sulfate (SO₄):
    ≤2 ppm
  • Trace Impurities - Aluminum (Al):
    ≤0.300 ppm
  • Trace Impurities - Boron (B):
    ≤0.100 ppm
  • Trace Impurities - Calcium (Ca):
    ≤0.300 ppm
  • Trace Impurities - Chromium (Cr):
    ≤0.200 ppm
  • Trace Impurities - Copper (Cu):
    ≤0.10 ppm
  • Trace Impurities - Gold (Au):
    ≤0.300 ppm
  • Trace Impurities - Iron (Fe):
    ≤0.2 ppm
  • Trace Impurities - Lead (Pb):
    ≤0.200 ppm
  • Trace Impurities - Magnesium (Mg):
    ≤0.300 ppm
  • Trace Impurities - Manganese (Mn):
    ≤0.200 ppm
  • Trace Impurities - Nickel (Ni):
    ≤0.1 ppm
  • Trace Impurities - Potassium (K):
    ≤0.300 ppm
  • Trace Impurities - Sodium (Na):
    ≤2.000 ppm
  • Trace Impurities - Tin (Sn):
    ≤0.300 ppm
  • Trace Impurities - Titanium (Ti):
    ≤0.300 ppm
  • Trace Impurities - Zinc (Zn):
    ≤0.300 ppm

Specifications

  • Pack type:
    Drum, steel/PE outer
  • Size:
    385 lbs.

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.