MIR Performance Validation Kit, PerkinElmer
Mid Infrared IPV kit for Spectrum 10 Software, —
Catalog # 10133-738
Supplier: PerkinElmer
Supplier #: L1250404
Jump to:
Product Details & Documents
Instrument performance validation kits are available for both Mid-IR and Near-IR FT-IR instruments
Upgrade kits are available to replace the polystyrene TRM once the validity period has expired.
Delivery information: Kits include a polystyrene Traceable Reference Material (TRM) for abscissa calibration valid for three years, NG11 Schott Glass Reference Material (RM).
Specifications
- Description:Mid Infrared IPV kit for Spectrum 10 Software
- Thickness:—