VWR® Glove Liners
Full Finger Liners, Extra Thick, with Red Cuff, X-Large, White
Catalog # 10124-750
Product available on GSA Advantage®
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
LINER GLOVE AMBIDEXTROUS E-LR 20PAIRS Catalog #32935-587
Product 32935-587 is similar to 10124-750 but manufacturer and specifications may vary.
Product Details & Documents
Ergonomically designed glove liners wick away moisture for comfortable use with disposable gloves.
- Ambidextrous glove liners
- Reduces perspiration and wicks away moisture
- Low-lint, 100% 15 denier nylon composition
- Sturdy, launderable, and reusable
- Color-coded cuffs indicate size
- Pre-laundered; will not shrink
Liners provide protection and comfort when used with latex, nitrile, and PVC gloves. Fully launderable for reusability and economy.
Packaging: Glove Liners are packaged 20 pair per bag, 15 bags per case. They are packed in clean polyethylene bags that are free of silicone, dioctyl phthalate (DOP), and amides.
Specifications
- Glove color:White
- Glove size:X-Large
- Description:Full Finger Liners, Extra Thick, with Red Cuff
- Cat. No.:10124-750
- Cuff Style:Knit Wrist
- Ambidextrous/Hand-specific:Ambidextrous
- Disposable/Reusable:Reusable