Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
IODEX AR/18 - 15 GALLON DRUM
Catalog # 10121-212
Supplier:
Quip Laboratories
SDS
Expand image
IODEX AR/18 - 15 GALLON DRUM
Catalog # 10121-212
Supplier:
Quip Laboratories
Supplier Number:
IAR18-15GAL
SDS
Related Information
Documentation
SDS
ghs_IAR18.pdf
Documentation