Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ACIDULATE 45T - 5 GALLON CONTAINER
Catalog # 10121-150
Supplier:
Quip Laboratories
SDS
Expand image
ACIDULATE 45T - 5 GALLON CONTAINER
Catalog # 10121-150
Supplier:
Quip Laboratories
Supplier Number:
AD45T-5GAL
SDS
Related Information
Documentation
SDS
ghs_AD45T.pdf
Documentation