Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Microscope Slide Boxes

Microscope Slide Boxes

Microscope slide box, lining: cork, lock type: thumb-latch, 50 Slides, 209×86×35 mm, White
Catalog # 490015-112
Supplier:   Heathrow Scientific
Supplier #:  HS15996C
Microscope Slide Boxes
Catalog # 490015-112
Microscope Slide Boxes
Microscope Slide Boxes

Microscope slide box, lining: cork, lock type: thumb-latch, 50 Slides, 209×86×35 mm, White

Price
$31.06
Log in for availability
Each Case of 72
Product Details & Documents

Heavy-duty, two-piece microscope slide boxes are ideal for storing or transportating standard slides, 75×25 mm (3×1").

  • Designed to securely store slides in grooved slots
  • Molded feet secure stacking for easy transport and storage

Closes securely with a rust-resistant nickel-plated clasp and pin hinges. ABS plastic boxes available with either cork or foam lining.

Documents

Specifications
  • Capacity:
    50 Slides
  • Color:
    White
  • Description:
    Microscope slide box, lining: cork, lock type: thumb-latch
  • Dimensions:
    209×86×35 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.