Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Microscope Slide Boxes

Microscope Slide Boxes

Microscope slide box, lining: cork, lock type: thumb-latch, 50 Slides, 209×86×35 mm, Red
  490015-110
Supplier #:  HS15996B
Microscope Slide Boxes
  490015-110
Microscope Slide Boxes
Microscope Slide Boxes

Microscope slide box, lining: cork, lock type: thumb-latch, 50 Slides, 209×86×35 mm, Red

 
$23.32
   
Each Case of 72
 

Heavy-duty, two-piece microscope slide boxes are ideal for storing or transportating standard slides, 75×25 mm (3×1").

  • Designed to securely store slides in grooved slots
  • Molded feet secure stacking for easy transport and storage

Closes securely with a rust-resistant nickel-plated clasp and pin hinges. ABS plastic boxes available with either cork or foam lining.

Documents

 
  • Capacity:
    50 Slides
  • Color:
    Red
  • Description:
    Microscope slide box, lining: cork, lock type: thumb-latch
  • Dimensions:
    209×86×35 mm