Orders LinkContactUsLinkComponent
VWR® Clip-It Timer™ Timer
VWR® Clip-It Timer™ Timer
Catalog # 490012-774
VWR® Clip-It Timer™ Timer
Catalog # 490012-774
Supplier:  
VWR® Clip-It Timer™ Timer
VWR® Clip-It Timer™ Timer
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
VWR TIMER CLIP-IT Catalog #62379-606
Product 62379-606 is a replacement for 490012-774 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Pocket-size timer performs countdown, alarm, and stopwatch functions and can be used to catch lab test results, signal drying/incubating times, or as alarm for appointments.
  • Clips onto lab coat, notebook, or clipboard
  • Traceable®
  • Housed in a rugged, chemical-resistant ABS case
Features quartz-crystal accuracy and easy-to-use three-button operation. Four-digit 8 mm (⁵/₁₆") high LCD displays time remaining in hours and minutes. Countdown mode programmable from 100 hours to 1 second. Stopwatch mode times from 1 second to 99 hours and then repeats itself. Timer also has time-out feature in both up and down counting. Loud beeping alarm sounds for 60 seconds.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with magnet for placing on metal and an 18-month continuous-use, 1.5 V silver-oxide battery.