Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Microscope Slide Boxes

Microscope Slide Boxes

Microscope slide box, lining: cork, lock type: nickel-plated clasp, 100 Slides, 208×175×34 mm, Red
Catalog # 490012-618
Supplier:   Heathrow Scientific
Supplier #:  HS15994C
Microscope Slide Boxes
Catalog # 490012-618
Microscope Slide Boxes
Microscope Slide Boxes

Microscope slide box, lining: cork, lock type: nickel-plated clasp, 100 Slides, 208×175×34 mm, Red

Price
$22.06
Log in for availability
Each Case of 72
Product Details & Documents

Heavy-duty, two-piece microscope slide boxes are ideal for storing or transportating standard slides, 75×25 mm (3×1").

  • Designed to securely store slides in grooved slots
  • Molded feet secure stacking for easy transport and storage

Closes securely with a rust-resistant nickel-plated clasp and pin hinges. ABS plastic boxes available with either cork or foam lining.

Documents

Specifications
  • Capacity:
    100 Slides
  • Color:
    Red
  • Description:
    Microscope slide box, lining: cork, lock type: nickel-plated clasp
  • Dimensions:
    208×175×34 mm