Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Cooling Containers

Cooling Containers

Pan, 1 L, Green
Catalog # 490007-648
Supplier:   Heathrow Scientific
Supplier #:  HS28721G
Cooling Containers
Catalog # 490007-648
Cooling Containers
Cooling Containers

Pan, 1 L, Green

Price
$123.32
Log in for availability
Each Case of 6
Product Details & Documents

Durable containers for chilling temperature-sensitive samples.

  • Made from polyurethane with metal inserts for stability
  • Close-fit lid keeps evaporation to a minimum and features a comfortable recessed knob
  • Tapered design for easy stacking
  • Precise pour spout and large handles
  • The lid and bottom of the bucket/pan are keyed to fit together
  • For low-temperature use to −80 °C

Suitable for low temperature use. Ideal for ice, ice slurries and CO₂ pellets.

Specifications
  • Capacity:
    1 L
  • Color:
    Green
  • Description:
    Pan
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.