Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Cooling Containers

Cooling Containers

Bucket, 4 L, Black
Catalog # 490007-646
Supplier:   Heathrow Scientific
Supplier #:  HS28714K
Cooling Containers
Catalog # 490007-646
Cooling Containers
Cooling Containers

Bucket, 4 L, Black

Price
$338.62
Log in for availability
Each Case of 6
Product Details & Documents

Durable containers for chilling temperature-sensitive samples.

  • Made from polyurethane with metal inserts for stability
  • Close-fit lid keeps evaporation to a minimum and features a comfortable recessed knob
  • Tapered design for easy stacking
  • Precise pour spout and large handles
  • The lid and bottom of the bucket/pan are keyed to fit together
  • For low-temperature use to −80 °C

Suitable for low temperature use. Ideal for ice, ice slurries and CO₂ pellets.

Specifications
  • Capacity:
    4 L
  • Color:
    Black
  • Description:
    Bucket