Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Advansta Buffer Pouches

Advansta Buffer Pouches

Buffer TBS Pouches
Catalog # 490005-046
Supplier:   Advansta
Supplier #:  R-01039-020
Advansta Buffer Pouches
Catalog # 490005-046
Supplier:  Advansta
CAS Number:  
Advansta Buffer Pouches
Advansta Buffer Pouches

Buffer TBS Pouches

CAS Number:  
Price
$137.85
Log in for availability
Pack of 20
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Convenient, pure, fast buffer preparation

  • Quality- Molecular biology-grade buffers of highest purity
  • Convenient- Pre-measured powder in sealed pouches
  • Fast- Dissolve and use right away
  • Standardized- Guaranteed reproducible results for your electrophoresis gels and Western blots

Advansta's Avant buffer pouches are perfect for standardizing your electrophoresis and Western blotting applications. Dissolve a pouch of pre-messured molecular biology-grade chemicals in 500 mL of deionized water to obtain a ready-to-use 1X buffer without any further preparation. Save precious time in the laboratory and generate reproducible results with Avant buffers.

Specifications

Specifications

  • Description:
    Buffer TBS Pouches

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.