Conductivity Indicator
Bulb (15W) for Water Conductivity
Catalog # 470061-754
Supplier: Science First
Supplier #: 674-0035
Jump to:
Product Details & Documents
Light Bulb Glows When Water Conducts Electricity.
- Attaches to a standard ring stand
- Comes with illustrated instructions
Presenting no shock hazard because the cord at the side can’t come in contact with the water, this indicator features a plastic molded body and recessed electrical connections and electrodes. It operates on 110 VAC. A light bulb is required.
Specifications
- Description:Bulb (15W) for Water Conductivity
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



