Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Charts and Pens for 6" Dickson® Chart Recorders, Graphic Controls

Chart Number 656, 24 h, 50...100 °C/°F
Catalog # 11032-018
Supplier:   Graphic Controls
 :  32020506
Charts and Pens for 6" Dickson® Chart Recorders, Graphic Controls
Catalog # 11032-018
Supplier:  Graphic Controls
Charts and Pens for 6" Dickson® Chart Recorders, Graphic Controls
Charts and Pens for 6" Dickson® Chart Recorders, Graphic Controls

Chart Number 656, 24 h, 50...100 °C/°F

Price
$132.84
Log in for availability
Pack of 60
Product Details & Documents

Graphic Controls supplies a full line of charts and markers to fit all available Dickson® recorders

Graphic Controls charts and markers provide precise fit and function for all available Dickson® recorders. Our products deliver OEM performance at factory direct savings.

Versatile and durable, Dickson® recorders are used in a wide variety of applications and conditions measuring temperature, humidity, and pressure. Graphic Controls charts and markers perfectly complement these diverse applications and conditions to ensure your critical data is accurately recorded and preserved.

 : For information on additional charts from Graphic Controls, please contact your Avantor sales representative.

Specifications
  • Temperature range:
    50...100 °C/°F
  • Description:
    Chart Number 656
  • Recording Time:
    24 h
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Blue Markers/Pens, 226
    Blue Markers/Pens, 226
    11032-080
    Log in for availability
    Pack of 6 - $199.39

Product Information

  • Catalog #
    Availability
    Price
  • Three Red & Three Blue Markers/Pens, 246
    Three Red & Three Blue Markers/Pens, 246
    11032-076
    Log in for availability
    Pack of 6 - $126.73

Product Information

  • Catalog #
    Availability
    Price
  • Red Markers/Pens, 222
    Red Markers/Pens, 222
    11032-078
    Log in for availability
    Pack of 6 - $118.41