Dilution Bottles
Dilution bottle, Phosphate buffer Magnesium Chloride, 99 ml
Supplier: GVS FILTER TECHNOLOGY
Supplier #: 10498505
Jump to:
Product Details & Documents
GVS dilution bottles are pre-filled and sterile, available in 90 and 99 ml sizes.
- Pre-filled for easy handling
- Offered in 90 and 99 ml volumes for easy 1:10 and 1:100 dilutions
- Compliance with standard methods
GVS pre-filled sterile dilution bottles are designed for sample dilution of water, dairy products, foods, and pharmaceuticals prior to microbiological testing. Final pH for all solutions is 7.2 pH (±0.2 pH) at 25 °C. They come in an easy open, flip-top, plastic container with a tamper-evident seal.
Documents
Specifications
- Capacity:99 ml
- Description:Dilution bottle, Phosphate buffer Magnesium Chloride
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



