GBS Detect/Strep Reagents
PYR Test Kit w/20ml Chromogenic Developer, —, 100 Tests
: Z175
GBS Detect is used for the isolation and differentiation by enhanced hemolysis of gamma hemolytic Group B Streptococcus.
- Grouping reagents for the rapid latex agglutination test
- Lancefield grouping includes A,B,C,D,F,G latex, controls, extraction reagents, cards, and mixing sticks
- StrepQuick™ to aid in the identification of gram positive cocci
GBS Detect™, for non-hemolytic Group B Strep. Granada Medium for hemolytic Group B Strep. Grouping Reagents A, B, C, and G, for the rapid latex agglutination test. PYR Test Kit with chromogenic developer on paper disks. Selective Beta Strep Agar, for Group A Strep only (Streptococcus pyogenes), especially for throat cultures. Selective Strep Agar (COBA Medium), for the selective isolation of all species of Strep. ci. StrepQuick™ performs a PYR, LAP, and Esculin test to aid in the identification of gram positive coc.
- Description:PYR Test Kit w/20ml Chromogenic Developer
- Size:100 Tests
- Format:—