Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CIDEHOL 70 5GL
Catalog # CI89125-212ABV
Supplier:
Decon Labs
SDS
Certificates
Expand image
CIDEHOL 70 5GL
Catalog # CI89125-212ABV
Supplier:
Decon Labs
Supplier Number:
8406
SDS
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
SDS
ghs_8406
TX89125-212ABV_A07152012B
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation