WHEATON® MicroLiter Inserts for Vials, DWK Life Sciences
Insert, 0.30 ml, Flat bottom, No
Catalog # 10039-404
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: 08-0000-150
WHEATON® MicroLiter Inserts for Vials, DWK Life Sciences
Catalog # 10039-404
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
Product Details & Documents
Microliter can help you determine the best products for your application.
- High-quality inserts for vials
Microliter has a full line of LC and GC Chromatography consumables and can help you determine the best products for your application.
Packaging: Conveniently packaged in packs of 100.
Documents
Specifications
- Cap included:No
- Capacity:0.30 ml
- Description:Insert
- Bottom style:Flat bottom