Lead(II) oxide, EMSURE® for analysis, Supelco®
Plastic bottle, 250 g
Supplier: MilliporeSigma
Supplier #: 1074010250
Lead(II) oxide, EMSURE® for analysis, Supelco®
Catalog # EM1.07401.0250
Supplier: MilliporeSigma
CAS Number: 1317-36-8
Jump to:
Product Details & Documents
Synonyms: Lead monoxide
- CAS Number:1317-36-8
- Formula:PbO
- MDL Number:MFCD00011164
- Molecular Weight:223.2 g/mol
- UN:3077
- ADR:9,III
Documents
Certificates
Search by Lot Numberghs_1.07401
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specification Test Results
- Solubility:0.017 g/l (20 °C)
- Melting Point:888 °C
- Molar Mass:223.19 g/mol
- Bulk Density:3500 - 3700 kg/m3
- Boiling Point:1470 °C
- Density:9.53 g/cm3 (20 °C)
- pH:8 - 9 (100 g/l, H2O, 20 °C) (slurry)
Specifications
- Pack type:Plastic bottle
- Size:250 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



