Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PH 110 METER KIT
Catalog # 101076-436
Supplier:
OAKTON INSTRUMENTS MS
Expand image
PH 110 METER KIT
Catalog # 101076-436
Supplier:
OAKTON INSTRUMENTS MS
Supplier Number:
WD-35615-80