$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
PH CON/10 METER KIT
101076-376
:
OAKTON INSTRUMENTS MS
PH CON/10 METER KIT
101076-376
:
OAKTON INSTRUMENTS MS
:
WD-35630-62