iMark™ Microplate Absorbance Reader, Bio-Rad
Product Details & Documents
The iMark™ Microplate Absorbance Reader, with a wavelength range of 400–750 nm, is an economical solution for a wide range of applications, including immunoassays with colorimetric substrates, such as ELISA and protein assays
- Fast microplate reading speeds
- Convenient built-in shaker
- Internal wide-format thermal graphical printer
- Easy-to-use, stand-alone operation
- Extensive onboard assay configuration
- Onboard software — includes programmable kinetic reading, comprehensive curve-fit selections
- Security login that allows locking and unlocking of key protocol settings by the administrator
- Checkmark™ Reader Performance Verification Kit has preprogrammed protocols to validate the IQ/OQ/PQ in compliance with GLP
- Ordering information: The iMark™ Microplate Absorbance Reader includes 415, 450, 490, 595, 655, and 750 nm filters, 8-filter capacity filter wheel, built-in plate shaker, onboard software and thermal printer, one roll printer paper, USB, and power cables. Microplate Manager® 6 Software is included with 1681135. A large selection of additional absorbance filters, mostly at 5 nm intervals from 400 to 700 nm, are available (see Accessories tab). The Model 1575 Immunowash™ Microplate Washer includes microplate washer, bottle, tubing, 8-port manifold, and aerosol protection cover.
The instrument can be used as a stand-alone reader using the 64 built-in assay protocols, data storage, analysis, and reports. The Microplate Manager® Software (PC or Mac) can be used to control the reader and format readings, plus more extensive data analysis.
Specifications
- Description:iMark™ Microplate Absorbance Reader
- Duration:0 - 999 s
- Languages:4 languages (English, Chinese, Japanese, Russian); LCD indication supported; printout report supported
- Time:6 sec at single wavelength, 10 sec at dual wavelengths
- Range:0.0–3.5 OD
- Printer:Built-in, thermal
- Wavelength:400–750 nm
- Weight:5.5 kg (12 lbs.)
- Cat. No.:100447-556
- Accuracy:≤1.0% or 0.010 from 0.000–3.000 OD at 490 nm
- Filters:415, 450, 490, 595, 655, and 750 nm
- Precision:1.0% or 0.005 OD from 0.0–2.0 OD; 1.5% from 2.0–3.0 OD
- Linearity:≤1.0% from 0.0–2.0 OD; ≤2.0% from 0.0–3.0 OD
- Voltage:100/240 V
- Output:Onboard graphical thermal printer and USB2 interface with PC or Mac data stations
- W×D×H:34.6×37.7×16.4 cm (13.6×14.8×6.5")
- Shaking Speed:Low, mid, high
- Data Storage:Calender/clock functions; 64 assay protocols
- Resolution:0.001 OD
Accessories
Product Information
- Catalog #AvailabilityPrice
- Replacement Thermal Paper, 3 Rolls
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



