Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Benzonase®

Benzonase®

100000 U, Plastic bottle
Catalog # EM1.01654.0001
Supplier #:  1.01654.0001
Benzonase®
Catalog # EM1.01654.0001
Supplier:  
CAS Number:  
Benzonase®

100000 U, Plastic bottle

CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Suitable for biopharmaceutical production EMPROVE® bio.

  • Activity (DNA; pH 8.0; 30 min; 37 °C) : ≥250
  • U/µL
  • Proteases: not detectable
  • Endotoxines (LAL-test): <0.25 EU in 1000 U
  • Colony count (aerobic bacteria), (Yeasts and moulds): <10 CFU in 100 000 U, <10 CFU in 100 000 U
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100000 U
  • Storage Conditions:
    Store at −20 °C. Transport with dry ice
  • Cat. No.:
    EM1.01654.0001
  • Form:
    Colorless
  • Purity:
    ≥99.0%(calc. based on protein), Grade II
  • Enzyme Activity:
    ≥1.1E+06 U/mg
  • Enzyme Name:
    Benzonase®

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.