Remove-iT Endo S
6000 units
Catalog # 102855-180
Small Business Enterprise
Supplier: New England Biolabs (NEB)
Supplier #: P0741S
Remove-iT Endo S
Catalog # 102855-180
Supplier: New England Biolabs (NEB)
Small Business Enterprise
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Remove-iT® Endo S is an endoglycosidase with a uniquely high specificity for removing N-linked glycans from the chitobiose core of the heavy chain of native IgG
Remove-iT Endo S is tagged with a chitin binding domain (CBD) for easy removal from a reaction and is supplied glycerol free for optimal performance in HPLC and MS intensive methods.
Specifications
Specifications
- Size:6000 units
- Cat. No.:102855-180
- Enzyme Name:Remove-iT Endo S