Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
PYREX® Volumetric Flasks

PYREX® Volumetric Flasks

50 ml
Catalog # 470211-518
   Low Manufacturing Impact
Supplier:   Corning
Supplier #:  5580-50
PYREX® Volumetric Flasks
Catalog # 470211-518
Supplier:  Corning
   Low Manufacturing Impact
PYREX® Volumetric Flasks
PYREX® Volumetric Flasks

50 ml

Price
$49.30
Log in for availability
Each Case of 12
Product Details & Documents

PYREX® glass, clear, with snap-caps.

  • Heavy beads and tubing rings tooled for snap-cap fit
  • Graduation line is sharp and permanent
  • No snap caps for 1 and 2 ml sizes
Specifications
  • Capacity:
    50 ml
  • Cat. No.:
    470211-518
  • Environmental Certification:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.