Orders LinkContactUsLinkComponent
HemaType Segment Device
HemaType segment device
Catalog # 89426-272
Supplier:   Decon Labs
 :  2000
HemaType Segment Device
Catalog # 89426-272
Supplier:  Decon Labs
HemaType Segment Device
HemaType Segment Device
HemaType segment device
Price
$672.69
Log in for availability
Pack of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Hematype Segment Devices are used for obtaining blood samples from blood bag tubing segments.
  • Easy-to-use
  • Reduces risk of blood spill
  • Single-use device
Decon's Hematype Segment device supports safe handling of blood bag tubing segments.
Specifications
  • Description:
    HemaType segment device