Ethidium bromide ≥95.0%
Product Details & Documents
CAS Number: 1239-45-8
MDL Number: MFCD00011724
Molecular Formula: C21H20BrN3
Molecular Weight: 394.32
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Flash Point (°C): 100
Product Categories: Bioscience, Stains & Dyes, Fluorescent Stains, Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium Bromide, Homidium Bromide
- CAS Number:1239-45-8
- Formula:C₂₁H₂₀BrN₃
- MDL Number:MFCD00011724
- Molecular Weight:394.31 g/mol
- Flash Pt:100 °C
- UN:2811
- ADR:6.1,I
Specifications
Specification Test Results
- Purity/Analysis Method:>95.0% (HPLC,T)
Specifications
- Size:1 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



