Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning

PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning

250 ml, 5
Catalog # 470202-162
Supplier:   Corning
Supplier #:  4060-250
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
Catalog # 470202-162
Supplier:  Corning
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning

250 ml, 5

Price
$28.10
Log in for availability
Each Case of 24
Product Details & Documents

Manufactured from borosilicate glass.

Tooled mouth ensures uniform stopper fit. Florence, long neck. Normal working temperature is 230 °C (446 °F).

Documents

Specifications
  • Capacity:
    250 ml
  • Stopper no.:
    5
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.