Order Entry
Puerto Rico
ContactUsLinkComponent
BATH TANK 19A PLEXI GL 55X33/17CM 19L
Catalog # 89422-028
Supplier:  JULABO
undefined
BATH TANK 19A PLEXI GL 55X33/17CM 19L
Catalog # 89422-028
Supplier:  JULABO
Supplier Number:  9900319