Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Violet
Catalog # 73960-417
Supplier: Simport Scientific
: T312-13
Capinsert™ Color Coding Inserts for Cryovial® Tubes, Simport Scientific
Catalog # 73960-417
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Color coded inserts fit precisely into the cap of the Cryovial® tubes for color identification.
Made of polypropylene
Ordering information: T312-7 contains 100 pieces each of white, blue, red, green and yellow inserts packaged in individual bags.
Documents
Specifications
- Color:Violet