Orders LinkContactUsLinkComponent
VWR® Florence Boiling Flasks
VWR® Florence Boiling Flasks
1000 ml
Catalog # 470191-210
Supplier:   VWR International
VWR® Florence Boiling Flasks
Catalog # 470191-210
Supplier:  VWR International
VWR® Florence Boiling Flasks
VWR® Florence Boiling Flasks
1000 ml
Price
$28.00
Log in for availability
Each Case of 6
Product Details & Documents
Our high-quality borosilicate Florence flasks meet ASTM standards and are a great value.
  • Boil liquids without excessive rapid evaporation
  • High-quality borosilicate glass fabrication
Their round body and long neck are ideal for boiling liquids without excessively rapid evaporation.
Documents
Specifications
  • Capacity:
    1000 ml