Orders LinkContactUsLinkComponent
Ward's® Basalt (Vesicular)
Basalt (Vesicular), bulk pack, 1 kg, Size will vary
Catalog # 470025-926
Supplier:   Avantor
Ward's® Basalt (Vesicular)
Catalog # 470025-926
Supplier:  Avantor
Ward's® Basalt (Vesicular)
Ward's® Basalt (Vesicular)
Basalt (Vesicular), bulk pack, 1 kg, Size will vary
Price
$22.79
Log in for availability
Each
Product Details & Documents
Dark gray with abundant gas pocket vesicles.
  • Collected in California
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" - 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Specifications
  • Description:
    Basalt (Vesicular), bulk pack, 1 kg
  • Specimen dimensions:
    Size will vary