Orders LinkContactUsLinkComponent
Tuning Fork Set
Tuning Fork Set
Tuning Forks, Set of 13 with Mallet
Catalog # 470102-472
Supplier #:  TFBOX13
Tuning Fork Set
Catalog # 470102-472
Tuning Fork Set
Tuning Fork Set
Tuning Forks, Set of 13 with Mallet
Clearance
$114.00
Sale
Valid until 31/12/2026
Log in for availability
Each Kit of 1
Product Details & Documents
Both versions of these economy tuning fork sets come with a mallet designed for one hard and one soft end for use with all frequencies of forks.
  • NAS Aluminum Alloy Construction
  • Mallet Included
  • Wooden Box with Die-Cut Foam Lining Provides Easy Storage
  • Two Sets Available
Both sets also come in a wooden box with die-cut foam lining for easy storage. All tuning forks are made of NAS aluminum alloy.
One set includes eight tuning forks (256, 288, 320, 341.3, 384, 426.6, 480, and 512). The other includes 13 tuning forks (100, 128, 256, 288, 320, 341.3, 384, 426.6, 480, 512, 1024, 2048, and 4096).
Specifications
  • Description:
    Tuning Forks, Set of 13 with Mallet