Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Banana Plug Patch Cords

Banana Plug Patch Cords

Patch cord, black, 61 cm, banana plugs both ends
Catalog # 470176-242
Supplier #:  1350302150001
Banana Plug Patch Cords
Catalog # 470176-242
Supplier:  
Banana Plug Patch Cords
Banana Plug Patch Cords

Patch cord, black, 61 cm, banana plugs both ends

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
ALLIGATOR /BANANA CORD BLACK 61 CM Catalog #470189-212

Product 470189-212 is similar to 470176-242 but may have a different manufacturer and/or packaging.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Double ended banana plug jumper wires.

  • Available in red or black
  • 15 cm (6") or 61 cm (24") long

Stackable 4 mm banana plugs on both ends of these flexible, plastic coated wires. The wires have a rating of 15 A, AC.

Specifications
  • Description:
    Patch cord, black, 61 cm, banana plugs both ends

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.