Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Assorted Wires

Insulated copper wire, 4 oz., 30 m, 18
  470092-832
 :  T-18
Assorted Wires
  470092-832
Assorted Wires
Assorted Wires

Insulated copper wire, 4 oz., 30 m, 18

 
$29.65
   
Each
 
 
  • Description:
    Insulated copper wire, 4 oz.
  • Length:
    30 m
  • Gauge:
    18