Single Factor Water Test Kits
Product Details & Documents
Simply Dip in Water and Read the Results
These kits feature a wide range of test factors for a fraction of the price of other single factor test kits. These test kits also eliminate the need for liquid reagents, hazardous chemicals, and glassware. Each kit comes with instructions.
Specifications
- Description:Single factor water test kit, pH, 30 tests