Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
VWR® Lab Notebook

VWR® Lab Notebook

VWR® Lab Notebook, Gridded, Black, 100, 28.1 cm, 21.8 cm
Catalog # 470150-762
Supplier:   VWR International
VWR® Lab Notebook
Catalog # 470150-762
Supplier:  VWR International
VWR® Lab Notebook
VWR® Lab Notebook

VWR® Lab Notebook, Gridded, Black, 100, 28.1 cm, 21.8 cm

Price
$40.25
Log in for availability
Each Case of 12
Product Details & Documents

Keep track of lab data and results with this convenient lab notebook.

  • Available in two styles
  • Hardcover
  • 100 Pages

In addition to blank pages for student records and remarks, it includes basic instructions on how to properly do a lab and state the outcomes. A full page periodic chart of the elements and conversion charts are also included for easy access to reference information.

Specifications
  • Cover color:
    Black
  • Description:
    VWR® Lab Notebook, Gridded
  • Length:
    21.8 cm
  • No. of pages:
    100
  • Width:
    28.1 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.