VWR® Lab Notebook
VWR® Lab Notebook, Lined, Teal, 100, 28.1 cm, 21.8 cm
470150-610
Keep track of lab data and results with this convenient lab notebook.
- Available in two styles
- Hardcover
- 100 Pages
In addition to blank pages for student records and remarks, it includes basic instructions on how to properly do a lab and state the outcomes. A full page periodic chart of the elements and conversion charts are also included for easy access to reference information.
- Cover color:Teal
- Description:VWR® Lab Notebook, Lined
- Length:21.8 cm
- No. of pages:100
- Width:28.1 cm



