Bulb socket board
Five-Socket
Catalog # 470149-512
Supplier: Shiv Dial Sud & Sons
: PH90778
Jump to:
Product Details & Documents
This solid socket board holds light bulbs for circuit experiments.
- Choice of Three or Five-Socket Sizes
- Removable Shunt Wires Featured on Five-Socket Size
- Terminals Featured on Three-Socket Board
- Plastic Bases with Rubber Feet
- Small Incandescent Bulbs
It's available in three or five-socket sizes. Terminals on three-socket board let users connect lamps in series or parallel, and five-socket board features removable shunt wires for the same purpose.
All units have plastic bases with rubber feet, and small incandescent bulbs are included.
Specifications
- Description:Five-Socket