Orders LinkContactUsLinkComponent
Single Factor Water Test Kits
Single Factor Water Test Kits
Single factor water test kit, total hardness, 30 tests
Catalog # 470144-634
Supplier #:  481108
Single Factor Water Test Kits
Catalog # 470144-634
Single Factor Water Test Kits
Single Factor Water Test Kits
Single factor water test kit, total hardness, 30 tests
Price
$18.00
Log in for availability
Each Case of 12
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Simply Dip in Water and Read the Results
These kits feature a wide range of test factors for a fraction of the price of other single factor test kits. These test kits also eliminate the need for liquid reagents, hazardous chemicals, and glassware. Each kit comes with instructions.
Specifications
  • Description:
    Single factor water test kit, total hardness, 30 tests