Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Litmus Test Sheets

Litmus Test Sheets

Litmus Test Sheets, Litmus Red
Catalog # 470123-138
Supplier #:  290-24-8X10
Litmus Test Sheets
Catalog # 470123-138
Litmus Test Sheets
Litmus Test Sheets

Litmus Test Sheets, Litmus Red

Price
$42.60
Log in for availability
Each Pack of 24
Product Details & Documents

These litmus test sheets are available in litmus red (ACS) or litmus blue (ACS).

  • Individual Sheet Length: 8"
  • Individual Width: 10"
  • Twenty-Four Sheets in One Package
  • Litmus Blue and Litmus Red Available

These individual sheets of litmus test papers are 8 x 10".

One package has 24 sheets.

Packaging: One package has 24 sheets.

Specifications
  • Description:
    Litmus Test Sheets, Litmus Red
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.