Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Spherical Mirrors

Convex mirror, –250 mm, 38 mm
  470111-308
 :  MCV310
Spherical Mirrors
  470111-308
Spherical Mirrors
Spherical Mirrors

Convex mirror, –250 mm, 38 mm

 
$7.00
   
Each
 

These wide range of mirrors are perfect for investigating real and virtual images.

  • Variety of Sizes and Focal Lengths
  • Circular
  • Polished Glass Construction
  • Durable
  • Back-Surface Silvered

The circular mirrors are available in diameters of 38 mm, 50 mm, and 75 mm.

They're back-surface silvered, made of polished glass, and painted for durability.

 
  • Description:
    Convex mirror
  • Focal length:
    –250 mm
  • Diameter:
    38 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.