Spherical Mirrors
Convex mirror, –100 mm, 38 mm
470111-304
: MCV304
These wide range of mirrors are perfect for investigating real and virtual images.
- Variety of Sizes and Focal Lengths
- Circular
- Polished Glass Construction
- Durable
- Back-Surface Silvered
The circular mirrors are available in diameters of 38 mm, 50 mm, and 75 mm.
They're back-surface silvered, made of polished glass, and painted for durability.
- Description:Convex mirror
- Focal length:–100 mm
- Diameter:38 mm



