Orders LinkContactUsLinkComponent
Right Angle Prisms
Right Angle Prisms
Right Angle Prisms, Glass, 32 x 50 mm
Catalog # 470111-284
Supplier #:  PFG050
Right Angle Prisms
Catalog # 470111-284
Right Angle Prisms
Right Angle Prisms
Right Angle Prisms, Glass, 32 x 50 mm
Price
$18.60
Log in for availability
Each
Product Details & Documents
For Versatile Optical Applications
  • These prisms are polished on all faces
  • They are made of shatter-resistant acrylic material or optical quality glass
These prisms are polished on all faces. They are made of shatter-resistant acrylic material or optical quality glass. The length of the hypotenuse and the prism height are given for each item.
Specifications
  • Description:
    Right Angle Prisms, Glass, 32 x 50 mm