Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Medium Grade, 15 μm

Filter Paper, Medium Grade, 110 mm diameter
  470019-884
 :  FPR011
Medium Grade, 15 μm
  470019-884
Medium Grade, 15 μm
Medium Grade, 15 μm

Filter Paper, Medium Grade, 110 mm diameter

 
$9.80
   
Box of 100 Each
 

The most widely used qualitative filter paper for routine applications.

  • Medium grade, 15 μm

Holds flocculants and precipitates commonly encountered in laboratory exercises.

 : Packages of 100.

 
  • Description:
    Filter Paper, Medium Grade, 110 mm diameter