Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Bone Cutting Shears

Bone Cutting Shears

Bone Cutting Shears, 222 mm (8.75"), Stainless steel
Catalog # 470018-972
Supplier:   Dunrite Instruments
Supplier #:  141110
Bone Cutting Shears
Catalog # 470018-972
Bone Cutting Shears
Bone Cutting Shears

Bone Cutting Shears, 222 mm (8.75"), Stainless steel

Price
$58.40
Log in for availability
Each
Product Details & Documents

Makes easy work of cutting bone.

  • Flush cutting face
  • Spring handles
  • Stainless steel construction wont chip
  • Stainless construction

Make one of the toughest dissection chores — cutting through bone — easier with straight shears designed especially for the job. These shears will also spare wear and tear on your fine instruments.

 : These instruments are intended for training and educational use only. These are not intended for human or animal procedures.

Specifications
  • Description:
    Bone Cutting Shears
  • Length:
    222 mm (8.75")
  • Material:
    Stainless steel