Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Spherical Mirrors

Spherical Mirrors

Concave mirror, 70 mm, 75 mm
Catalog # 470016-996
Supplier #:  MCC703
Spherical Mirrors
Catalog # 470016-996
Spherical Mirrors
Spherical Mirrors

Concave mirror, 70 mm, 75 mm

Price
$7.00
Log in for availability
Each
Product Details & Documents

These wide range of mirrors are perfect for investigating real and virtual images.

  • Variety of Sizes and Focal Lengths
  • Circular
  • Polished Glass Construction
  • Durable
  • Back-Surface Silvered

The circular mirrors are available in diameters of 38 mm, 50 mm, and 75 mm.

They're back-surface silvered, made of polished glass, and painted for durability.

Specifications
  • Description:
    Concave mirror
  • Focal length:
    70 mm
  • Diameter:
    75 mm