Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Spherical Mirrors

Spherical Mirrors

Convex mirror, –50 mm, 38 mm
Catalog # 470005-646
Supplier #:  MCV302
Spherical Mirrors
Catalog # 470005-646
Spherical Mirrors
Spherical Mirrors

Convex mirror, –50 mm, 38 mm

Price
$7.00
Log in for availability
Each
Product Details & Documents

These wide range of mirrors are perfect for investigating real and virtual images.

  • Variety of Sizes and Focal Lengths
  • Circular
  • Polished Glass Construction
  • Durable
  • Back-Surface Silvered

The circular mirrors are available in diameters of 38 mm, 50 mm, and 75 mm.

They're back-surface silvered, made of polished glass, and painted for durability.

Specifications
  • Description:
    Convex mirror
  • Focal length:
    –50 mm
  • Diameter:
    38 mm